missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetylamino-3-(2-chlorobenzoyl)-5-(2-carbomethoxyethyl)thiophene, TRC

Product Code. 30329037
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329037 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329037 Supplier Toronto Research Chemicals Supplier No. TRCB67908510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoacetylamino-3-(2-chlorobenzoyl)-5-(2-carbomethoxyethyl)thiophene
CAS 100827-79-0
Molecular Formula C17H15BrClNO4S
Quantity 10 mg
Synonym 5-[(2-Bromoacetyl)amino]-4-(2-chlorobenzoyl)-2-thiophenepropanoic Acid Methyl Ester, 5-[(Bromoacetyl)amino]-4-(2-chlorobenzoyl)-2-thiophenepropanoic Acid Methyl Ester
InChI Formula InChI=1S/C17H15BrClNO4S/c1-24-15(22)7-6-10-8-12(17(25-10)20-14(21)9-18)16(23)11-4-2-3-5-13(11)19/h2-5,8H,6-7,9H2,1H3,(H,20,21)
SMILES COC(=O)CCc1cc(C(=O)c2ccccc2Cl)c(NC(=O)CBr)s1
IUPAC Name methyl 3-[5-[(2-bromoacetyl)amino]-4-(2-chlorobenzoyl)thiophen-2-yl]propanoate
Molecular Weight (g/mol) 444.73
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.