missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetyl Bromide, TRC

Product Code. 30335182
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335182 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335182 Supplier Toronto Research Chemicals Supplier No. TRCB67915010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromoacetyl bromide
CAS 598-21-0
Molecular Formula C2 H2 Br2 O
Quantity 10 g
Synonym Bromoacetyl Bromide, Monobromoacetyl Bromide, α-Bromoacetyl Bromide
InChI Formula InChI=1S/C2H2Br2O/c3-1-2(4)5/h1H2
SMILES BrCC(=O)Br
IUPAC Name 2-bromoacetyl bromide
Molecular Weight (g/mol) 201.84
Formula Weight 199.8472
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.