missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetic Acid, TRC

Product Code. 30335730
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335730 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335730 Supplier Toronto Research Chemicals Supplier No. TRCB67907510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromoacetic acid
CAS 79-08-3
Purity Grade Notes GC
Molecular Formula C2 H3 Br O2
Quantity 10 g
Synonym Acetic acid, bromo- (8CI,9CI), 2-Bromoacetic acid, 2-Bromoethanoic acid, Bromoacetate ion, Bromoacetic acid, Bromoethanoic acid, Carboxymethyl bromide, MBAA, Monobromoacetic acid, NSC 141, To NTu, α-Bromoacetic acid
InChI Formula InChI=1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)
SMILES OC(=O)CBr
IUPAC Name 2-bromoacetic acid
Molecular Weight (g/mol) 138.95
Formula Weight 137.9316
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.