missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N-methylbenzylamine, 95%, Thermo Scientific™

Product Code. 15430059
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15430059

Brand: Thermo Scientific Alfa Aesar H63390.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-19-1
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD05262953
InChI Key TUADRPBKJHMHDH-UHFFFAOYSA-N
Synonym 2-bromobenzyl methylamine, n-methyl-2-bromobenzylamine, 2-bromo-n-methylbenzylamine, 1-2-bromophenyl-n-methylmethanamine, 2-bromophenyl methyl methyl amine, benzenemethanamine, 2-bromo-n-methyl, n-methyl-o-bromobenzylamine, acmc-20ao1g, 2-bromo-n-methyl benzylamine, 2-bromo-benzyl-methyl-amine
PubChem CID 522360
IUPAC Name 1-(2-bromophenyl)-N-methylmethanamine
SMILES CNCC1=CC=CC=C1Br

Specifications

Density 1.358 g/mL
Boiling Point 71°C to 74°C (2 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.567
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 200.08
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material 2-Bromo-N-methylbenzylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.