missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-alpha-methylbenzyl alcohol, 99%

Product Code. p-3598266 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10616254

Brand: Thermo Scientific 393640010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5411-56-3 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5411-56-3
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00065001
InChI Key DZLZSFZSPIUINR-UHFFFAOYSA-N
Synonym 1-2-bromophenyl ethanol, 2-bromo-alpha-methylbenzyl alcohol, 1-2-bromophenyl ethan-1-ol, 1-2'-bromophenyl-1-hydroxyethane, acmc-209wki, o-bromo-1-phenylethanol, o-bromo-1-phenyl ethanol, acmc-209y6n, 1-2-bro-mophenyl ethanol, 1-2-bromo-phenyl-ethanol
PubChem CID 95455
IUPAC Name 1-(2-bromophenyl)ethanol
SMILES CC(C1=CC=CC=C1Br)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 54°C to 56°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH(OH)CH3
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Quantity 1 g
Formula Weight 201.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromo-α-methylbenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-a-methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.