missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-fluorenone, 95%, ACROS Organics™

Product Code. 10346452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10346452 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10346452 Supplier Thermo Scientific Acros Supplier No. 354701000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3096-56-8
CAS Min % 94.0
CAS Max % 100.0
Melting Point 143°C to 149°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C13H7BrO
MDL Number MFCD00010789
Quantity 100 mg
Synonym 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f
InChI Key MTCARZDHUIEYMB-UHFFFAOYSA-N
SMILES BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1
IUPAC Name 2-bromo-9H-fluoren-9-one
Molecular Weight (g/mol) 259.10
PubChem CID 725831
Formula Weight 259.1
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material 2-Bromo-9-fluorenone, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.