missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-dimethylfluorene, 98%

Product Code. 15422039
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422039 5 g 5g
15432039 25 g 25g
15442039 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422039 Supplier Thermo Scientific Alfa Aesar Supplier No. H64732.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28320-31-2 | C15H13Br | 273.173 g/mol

This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28320-31-2
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD09261272
InChI Key MBHPOBSZPYEADG-UHFFFAOYSA-N
Synonym 2-bromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2-bromofluorene, 9h-fluorene, 2-bromo-9,9-dimethyl, 2-bromo-9,9-dimethyl-fluorene, 2-bromo-9,9-dimethylfluoren, ksc494q3r, 2-bromo-9,9-dimethyl fluorene, 9,9-dimethyl-2-bromofluorenone, 2-bromo-9,9-dimethylfluorene
PubChem CID 10945606
IUPAC Name 2-bromo-9,9-dimethylfluorene
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)C

Specifications

Melting Point 57°C to 62°C
Quantity 5 g
Beilstein 2524870
Solubility Information Slightly soluble in water.
Formula Weight 273.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-9,9-dimethylfluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.