missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-pyrrolidin-1-ylpyridine, ≥97%, Thermo Scientific™

Product Code. 10312305
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10312305 5 g 5g
10010213 250 mg 250mg
10706463 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10312305 Supplier Thermo Scientific Maybridge Supplier No. CC38010DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 230618-41-4
Molecular Formula C9H11BrN2
Molecular Weight (g/mol) 227.105
MDL Number MFCD08271892
InChI Key HZOWJDJNPISFJC-UHFFFAOYSA-N
Synonym 2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
PubChem CID 15470277
IUPAC Name 2-bromo-6-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=NC(=CC=C2)Br

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 227.1
Percent Purity 90%
Chemical Name or Material 2-Bromo-6-pyrrolidin-1-ylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.