missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-pyridinemethanol, 96%

Product Code. 15442558
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442558 5 g 5g
15452558 250 mg 250mg
15432558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442558 Supplier Thermo Scientific Alfa Aesar Supplier No. H56123.06
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 33674-96-3 | C6H6BrNO | 188.02 g/mol

It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and bases. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33674-96-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04039884
InChI Key XDDGKNRSCDEWBR-UHFFFAOYSA-N
Synonym 6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
PubChem CID 3517811
IUPAC Name (6-bromopyridin-2-yl)methanol
SMILES OCC1=CC=CC(Br)=N1

Specifications

Melting Point 34°C to 39°C
Boiling Point 246°C
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 188.02
Percent Purity 96%
Chemical Name or Material 2-Bromo-6-pyridinemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.