missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-piperidinopyridine, ≥97%, Thermo Scientific™

Product Code. 10259392
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10259392 1 g Amber Glass Bottle 1g
10269772 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10259392 Supplier Thermo Scientific Maybridge Supplier No. CC38110DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 24255-97-8
Molecular Formula C10H13BrN2
Molecular Weight (g/mol) 241.132
MDL Number MFCD07772823
InChI Key YHQZMCNKHWPFBS-UHFFFAOYSA-N
Synonym 2-bromo-6-piperidin-1-yl pyridine, 2-bromo-6-piperidinopyridine, acmc-1cmoy, 6-bromo-2-piperidylpyridine, 2-bromo-6-1-piperidinyl pyridine, 2-bromo-6-piperadinopyridine, 2-bromanyl-6-piperidin-1-yl-pyridine, pyridine,2-bromo-6-1-piperidinyl, 6'-bromo-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl
PubChem CID 7164597
IUPAC Name 2-bromo-6-piperidin-1-ylpyridine
SMILES C1CCN(CC1)C2=NC(=CC=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 241.13
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-piperidinopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.