missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-methyl-5-nitropyridine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-6-methyl-5-nitropyridine |
| CAS | 22282-96-8 |
| Molecular Formula | C6 H5 Br N2 O2 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C6H5BrN2O2/c1-4-5(9(10)11)2-3-6(7)8-4/h2-3H,1H3 |
| SMILES | Cc1nc(Br)ccc1[N+](=O)[O-] |
| IUPAC Name | 6-bromo-2-methyl-3-nitropyridine |
| Molecular Weight (g/mol) | 217.02 |
| Formula Weight | 215.9534 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction