missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxyphenol, 98+%

Product Code. 11481070
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481070 25 g 25g
11461070 1 g 1g
11471070 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481070 Supplier Thermo Scientific Alfa Aesar Supplier No. B25535.14
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 28165-49-3 | C7H7BrO2 | 203.035 g/mol

2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28165-49-3
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD08146628
InChI Key WEUFQISIJPSTBM-UHFFFAOYSA-N
PubChem CID 11019958
IUPAC Name 2-bromo-6-methoxyphenol
SMILES COC1=C(C(=CC=C1)Br)O

Specifications

Melting Point 62°C to 65°C
Quantity 25 g
Synonym 2-bromo-6-methoxy-phenol, phenol, 2-bromo-6-methoxy, bromhydroxyanisol, 6-bromoguaiacol, 3-bromo-2-hydroxyanisole, acmc-1ch0f, phenol,2-bromo-6-methoxy, 2-bromanyl-6-methoxy-phenol, ksc560s4p
Solubility Information Slightly soluble in water.
Formula Weight 203.04
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-6-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.