missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxynaphthalene (Naproxen Impurity N), TRC

Product Code. 30133121
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133121 10 g 10g
30132900 50 g 50g
17961005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133121 Supplier Toronto Research Chemicals Supplier No. TRCB68478010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5111-65-9
Molecular Formula C11 H9 Br O
Molecular Weight (g/mol) 237.09
Synonym 2-Bromo-6-methoxynaphthalene, Naproxen Imp. N (EP)
IUPAC Name 2-bromo-6-methoxynaphthalene
SMILES COc1ccc2cc(Br)ccc2c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H9BrO/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h2-7H,1H3
Formula Weight 235.9837
Percent Purity >95%
Chemical Name or Material 2-Bromo-6-methoxynaphthalene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.