missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-hydrazinopyridine, 98%, Thermo Scientific Chemicals

Product Code. 15461959
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26944-71-8
Molecular Formula C5H6BrN3
Molecular Weight (g/mol) 188.028
MDL Number MFCD09743754
InChI Key PQMFVUNERGGBPG-UHFFFAOYSA-N
Synonym 2-bromo-6-hydrazinylpyridine, 2-bromo-6-hydrazinopyridine, 2-6-bromopyridin-2-yl hydrazine, 6-bromo-pyridin-2-yl-hydrazine, 2-bromo-6-hydrazinopydine, 6-bromo-2-pyridyl hydrazine, 6-bromopyridin-2-yl hydrazine, 6-bromo-2-hydrazino pyridine, 2 1h-pyridinone, 6-bromo-, hydrazone, pubchem12987
PubChem CID 19392588
IUPAC Name (6-bromopyridin-2-yl)hydrazine
SMILES C1=CC(=NC(=C1)Br)NN

Specifications

Melting Point 117°C to 118°C
Color Brown
Quantity 1 g
UN Number UN2811
Beilstein 386966
Formula Weight 188.03
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-hydrazinopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'