missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoroaniline, 98+%

Product Code. p-7028451
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11424350

Brand: Thermo Scientific Alfa Aesar H25774.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65896-11-9 | C6H5BrFN | 190.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65896-11-9
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD01310982
InChI Key ALZFPYUPNVLVQM-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-fluoro, 2-bromo-6-fluoro-phenylamine, 2-bromo-6-fluorobenzenamine, 2-bromo-6-fluoro-aniline, 2-bromo-6-fluoro aniline, 2-bromo-6-fluorophenylamine, pubchem2923, acmc-209nsu, 2-bromo-6-flouroaniline, ksc493o8h
PubChem CID 2782940
IUPAC Name 2-bromo-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Br

Specifications

Density 1.674
Refractive Index 1.582
Quantity 5 g
UN Number UN2810
Beilstein 8541957
Formula Weight 190.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-6-fluoroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.