missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoroaniline, 98+%

Product Code. 11424350
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424350 5 g 5g
11434350 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424350 Supplier Thermo Scientific Alfa Aesar Supplier No. H25774.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65896-11-9 | C6H5BrFN | 190.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65896-11-9
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD01310982
InChI Key ALZFPYUPNVLVQM-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-fluoro, 2-bromo-6-fluoro-phenylamine, 2-bromo-6-fluorobenzenamine, 2-bromo-6-fluoro-aniline, 2-bromo-6-fluoro aniline, 2-bromo-6-fluorophenylamine, pubchem2923, acmc-209nsu, 2-bromo-6-flouroaniline, ksc493o8h
PubChem CID 2782940
IUPAC Name 2-bromo-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Br

Specifications

Density 1.674
Refractive Index 1.582
Quantity 5 g
UN Number UN2810
Beilstein 8541957
Formula Weight 190.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-6-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.