missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chlorobenzoic acid, 98%

Product Code. 15454977
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454977 1 g 1g
15464977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454977 Supplier Thermo Scientific Alfa Aesar Supplier No. H32269.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93224-85-2 | C7H4BrClO2 | 235.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93224-85-2
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00672929
InChI Key URGXUQODOUMRFP-UHFFFAOYSA-N
Synonym 6-bromo-2-chlorobenzoic acid, 2-chloro-6-bromobenzoic acid, benzoic acid, 2-bromo-6-chloro, 2-bromo-6-chloro-benzoic acid, buttpark 51\01-28, akos msc-0781, 2-bromo-6-chlorobenzoicacid, pubchem4048, acmc-209rkm, 2-chloro-6-bromobenzoic
PubChem CID 33125
IUPAC Name 2-bromo-6-chlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)C(=O)O)Cl

Specifications

Melting Point 148°C to 150°C
Quantity 1 g
Formula Weight 235.47
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-chlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.