missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile, 95%, Thermo Scientific Chemicals

Product Code. 15486816
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486816 250 mg 250mg
15476816 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486816 Supplier Thermo Scientific Alfa Aesar Supplier No. H61858.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1260763-01-6
Molecular Formula C12H14BrN3
Molecular Weight (g/mol) 280.169
InChI Key XDFWUVBVNHLPIB-UHFFFAOYSA-N
Synonym 2-bromo-6-4-methylpiperazin-1-yl benzonitrile
PubChem CID 75360874
IUPAC Name 2-bromo-6-(4-methylpiperazin-1-yl)benzonitrile
SMILES CN1CCN(CC1)C2=C(C(=CC=C2)Br)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 280.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.