missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)pyridine, 95+%, Maybridge™

Product Code. 10049541
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10049541 10 g Amber Glass Bottle 10g
10039541 1 g Amber Glass Bottle 1g
10124104 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10049541 Supplier Thermo Scientific Maybridge Supplier No. SPB05624EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50488-42-1
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
InChI Key GSKMWMFOQQBVMI-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl pyridine, 6-bromo-3-trifluoromethylpyridine, 2-bromo-5-trifluoromethyl-pyridine, pyridine, 2-bromo-5-trifluoromethyl, 5-trifluoromethyl-2-bromopyridine, 2-brom-5-trifluormethyl pyridin, 2-bromo-5-trilfuoromethylpyridine, pubchem3005
PubChem CID 2736434
IUPAC Name 2-bromo-5-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1C(F)(F)F)Br

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 225.99
Percent Purity 95%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.