missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11306569
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11306569 5 g 5g
11396559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306569 Supplier Thermo Scientific Alfa Aesar Supplier No. H26176.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176225-08-4
Molecular Formula C7H3BrClF3O2S
Molecular Weight (g/mol) 323.508
MDL Number MFCD03094395
InChI Key CBIFOAHDKKGDAC-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl benzenesulfonyl chloride, 2-bromo-5-trifluoromethyl benzene-1-sulfonyl chloride, 2-bromo-5-trifluoromethyl benzenesulfonylchloride, 2-bromo-5-trifluoromethyl benzenesulphonyl chloride, 4-bromo-3-chlorosulphonyl benzotrifluoride, 2-bromo-5-trifluoromethyl benzene sulfonyl chloride, pubchem11717, acmc-1c27d, 2-bromo-5-trifluoromethyl benzenesulphonylchloride, benzenesulfonylchloride,2-bromo-5-trifluoromethyl
PubChem CID 2778613
IUPAC Name 2-bromo-5-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)S(=O)(=O)Cl)Br

Specifications

Density 1.854
Boiling Point 230°C to 231°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 323.52
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.