missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)aniline, 97%

Product Code. 11403408
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403408 5 g 5g
11493398 1 g 1g
11413408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403408 Supplier Thermo Scientific Alfa Aesar Supplier No. A17810.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 454-79-5 | C7H5BrF3N | 240.023 g/mol

2-Bromo-5-(trifluoromethyl)aniline is used in the synthesis and biochemical evaluation of series of inhibitors of hepatitis C virus (HCV) NS3 protease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-(trifluoromethyl)aniline is used in the synthesis and biochemical evaluation of series of inhibitors of hepatitis C virus (HCV) NS3 protease.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-79-5
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
MDL Number MFCD00007637
InChI Key PZDVFXUBTKPFSG-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl aniline, 3-amino-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl benzenamine, 2-bromo-5-trifluoromethylphenylamine, 2-amino-1-bromo-4-trifluoromethylbenzene, 2-amino-4-trifluoromethylbromobenzene, 6-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, 3-amino-4-bromo trifluoromethyl benzene, benzenamine, 2-bromo-5-trifluoromethyl, 4-bromo-3-aminobenzotrifluoride
PubChem CID 67989
IUPAC Name 2-bromo-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)Br

Specifications

Density 1.675
Boiling Point 81°C to 82°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.522
Quantity 5 g
UN Number UN2810
Beilstein 2094535
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 240.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.