missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole, Thermo Scientific™

Product Code. 10381460
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10381460 5 g Amber Glass Bottle 5g
10595270 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10381460 Supplier Thermo Scientific Maybridge Supplier No. CC72310DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 37461-61-3
Molecular Formula C3BrF3N2S
Molecular Weight (g/mol) 233.01
MDL Number MFCD00015535
InChI Key LGTVLLPQCMJOGT-UHFFFAOYSA-N
PubChem CID 12874685
IUPAC Name 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole
SMILES FC(F)(F)C1=NN=C(Br)S1

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Synonym 2-bromo-5-trifluoromethyl-1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4 thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4-thiadiazol, 5-bromo-2-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazole,2-bromo-5-trifluoromethyl, 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl, 2-bromanyl-5-trifluoromethyl-1,3,4-thiadiazole
Formula Weight 233.01
Percent Purity 97%
Chemical Name or Material 2-bromo-5-(trifluoromethyl)-1, 3, 4-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.