missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiazole, 98%, Thermo Scientific™

Product Code. 10529820 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10529820 5 g Glass bottle 5g
10345930 1 g Glass bottle 1g
10289483 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10529820 Supplier Thermo Scientific Acros Supplier No. 107180050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-48-8
Molecular Formula C3HBrN2O2S
Molecular Weight (g/mol) 209.02
MDL Number MFCD00005317
InChI Key ANIJFZVZXZQFDH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, #
PubChem CID 18211
IUPAC Name 2-bromo-5-nitro-1,3-thiazole
SMILES [O-][N+](=O)C1=CN=C(Br)S1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 84°C to 88°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 209.02
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-5-nitrothiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.