missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiazole, 98%, Thermo Scientific™

Catalog Number 10345930
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3034-48-8
C3HBrN2O2S
209.02
MFCD00005317
ANIJFZVZXZQFDH-UHFFFAOYSA-N
2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, #
18211
2-bromo-5-nitro-1,3-thiazole
[O-][N+](=O)C1=CN=C(Br)S1
This item is not returnable. View return policy
3034-48-8
C3HBrN2O2S
209.02
MFCD00005317
ANIJFZVZXZQFDH-UHFFFAOYSA-N
2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, #
18211
2-bromo-5-nitro-1,3-thiazole
[O-][N+](=O)C1=CN=C(Br)S1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 84°C to 88°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 209.02
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-5-nitrothiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'