missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitropyridine, 98+%, Thermo Scientific™

Product Code. 11312737
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312737 1 g 1g
11322737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312737 Supplier Thermo Scientific Alfa Aesar Supplier No. L15661.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-5-nitropyridine was used in preparation of boc-protected (piperazin-1-ylmethyl)biaryls via microwave-mediated Suzuki-Miyaura coupling with (boc-piperazin-1-ylmethyl)phenylboronic acid pinacol esters. It was also used in the synthesis of 2-pyridyl analogs. Substituted 5-nitro-2-ethynylpyridines were synthesized by the Sonogashira reaction of 2-bromo-5-5-nitropyridine with terminal acetylenes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4487-59-6
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00006222
InChI Key HUUFTVUBFFESEN-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095
PubChem CID 78240
IUPAC Name 2-bromo-5-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Br)N=C1

Specifications

Melting Point 136°C to 140°C
Boiling Point 145°C to 147°C (10 mmHg)
Quantity 1 g
Beilstein 120901
Solubility Information Insoluble in water.
Formula Weight 203
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.