missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrobenzoic acid, 98%

Product Code. 11421737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421737 5 g 5g
11411737 1 g 1g
11339669 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421737 Supplier Thermo Scientific Alfa Aesar Supplier No. B25704.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 943-14-6 | C7H4BrNO4 | 246.016 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943-14-6
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00134558
InChI Key UVFWYVCDRKRAJH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179
PubChem CID 243025
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br

Specifications

Melting Point 178°C to 181°C
Quantity 5 g
Beilstein 980242
Formula Weight 246.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.