missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4%), 95%

Product Code. 10448278 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10448278 1 g Glass bottle 1g
10419238 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10448278 Supplier Thermo Scientific Supplier No. 434200010
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 765-58-2 | C5H5BrS | 177.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-58-2
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.06
MDL Number MFCD00130103
InChI Key ACDLOOGOFKSUPO-UHFFFAOYSA-N
Synonym 2-bromo-5-methyl-thiophene, 2-bromo-5-methyl thiophene, thiophene, 2-bromo-5-methyl, 2-methyl-5-bromothiophene, 2-brom-5-methylthiophen, zlchem 358, pubchem5193, acmc-1beki, 2-methyl-5-bromothiphene, 5-bromo-2-methylthiophene
PubChem CID 69831
IUPAC Name 2-bromo-5-methylthiophene
SMILES CC1=CC=C(S1)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Boiling Point 65°C (15.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Stabilizer stabilizer - Cu (0.1%), NaHCO3 (0.4%)
Formula Weight 177.06
Percent Purity ≥94.0%
Physical Form Liquid, May Include Precipitate
Chemical Name or Material 2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4%)
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4%), 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.