missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylbenzotrifluoride, Thermo Scientific™

Product Code. 10199409
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10199409 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10199409 Supplier Thermo Scientific Maybridge Supplier No. MAY00190DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 261952-20-9
Packaging Amber Glass Bottle
Molecular Formula C8H6BrF3
Quantity 1 g
Synonym 2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl
InChI Key WXXDBPIOFOYRLP-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)Br)C(F)(F)F
IUPAC Name 1-bromo-4-methyl-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 239.035
PubChem CID 2779339
Formula Weight 239.03
Percent Purity 95%
Chemical Name or Material 2-bromo-5-methylbenzotrifluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.