missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxytoluene, 97%, ACROS Organics™

Brand:  Acros Organics 294030050

Code : 29

Additional Details : CAS Number : 27060-75-9 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10407202

  • £27.00 / 5g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

2-Bromo-5-methoxytoluene
96.0
97%
C8H9BrO
MFCD00060687
27060-75-9
100.0
97%
201.06

Specifications

2-Bromo-5-methoxytoluene
96.0
Authentic
1.5600 to 1.5620
0.9100g/mL
5g
Clear colorless to slightly yellow
97%
C8H9BrO
MFCD00060687
27060-75-9
100.0
Glass bottle
0.91
111.0° to 112.0°C (15.0 mmHg)
>110°C
Liquid
97%
201.06
Videos
SDS