missing translation for 'onlineSavingsMsg'
Learn More

n-Butyllithium, 2.5M solution in hexanes, AcroSeal™

Product Code. 10030462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Glass bottle
Unit Size:
100mL
50mL
800mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10030462 100 mL AcroSeal™ Glass bottle 100mL
16243861 50 mL Glass bottle 50mL
10181852 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10030462 Supplier Thermo Scientific Acros Supplier No. 213351000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 109-72-8 | C4H9Li | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-72-8
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
PubChem CID 61028
SMILES [Li]CCCC

Specifications

Melting Point -95.0°C
Density 0.6900g/mL
Boiling Point 60.0°C to 80.0°C
Flash Point −21°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH3(CH2)3Li
Quantity 100 mL
Fieser 01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59
Specific Gravity 0.69
IUPAC Name lithium;butane
Formula Weight 64.06
Chemical Name or Material n-Butyllithium, 2.5M Solution in Hexanes
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butyllithium, 2.5M solution in hexanes
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Substance which, in contact with water, emits flammable gases Category 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.