missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxybenzaldehyde, 95%

Product Code. 15492279
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492279 100 g 100g
15472279 5 g 5g
15482279 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492279 Supplier Thermo Scientific Alfa Aesar Supplier No. H64884.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2973-80-0 | C7H5BrO2 | 201.02 g/mol

2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2973-80-0
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD03033065
InChI Key SCRQAWQJSSKCFN-UHFFFAOYSA-N
PubChem CID 387179
IUPAC Name 2-bromo-5-hydroxybenzaldehyde
SMILES OC1=CC(C=O)=C(Br)C=C1

Specifications

Melting Point 130°C to 135°C
Quantity 100 g
UN Number UN3077
Beilstein 2079921
Synonym 4-bromo-3-formylphenol, 2-bromo-5-hydroxy-benzaldehyde, 2-bromo-5-hydroxy benzaldehyde, benzaldehyde, 2-bromo-5-hydroxy, 2-bromo-5-hydroxybenzadehyde, benzaldehyde,2-bromo-5-hydroxy, pubchem14430, acmc-209haw, 6-brom-3-oxy-benzaldehyd
Solubility Information Soluble in chloroform,dichloromethane and ethyl acetate.
Formula Weight 201.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.