missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-5-fluorotoluene, 98+%

Brand:  Alfa Aesar™ A11112.14

Code : NEW

Additional Details : CAS Number : 452-63-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11478993

  • £38.20 / 25g


EU Stock 1
Add to basket

Description

Description

Applications
2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

2-Bromo-5-fluorotoluene
C7H6BrF
MFCD00017921
452-63-1
189.03
1859121

Specifications

2-Bromo-5-fluorotoluene
≥98%
25g
C7H6BrF
MFCD00017921
452-63-1
169° to 170°C
70°C (158°F)
189.03
1859121
Videos
SDS