missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorotoluene, 98+%

Product Code. 15493027
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493027 Supplier Thermo Scientific Alfa Aesar Supplier No. A11112.06
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 452-63-1 | C7H6BrF | 189.03 g/mol

2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 452-63-1
Density 1.495
Boiling Point 169°C to 170°C
Flash Point 70°C
Molecular Formula C7H6BrF
Refractive Index 1.526
MDL Number MFCD00017921
Quantity 5 g
Beilstein 1859121
Synonym 2-bromo-5-fluorotoluene, 3-fluoro-6-bromotoluene, 2-bromo-5-fluoro toluene, benzene, 1-bromo-4-fluoro-2-methyl, 5-fluoro-2-bromotoluene
Solubility Information Difficult to mix.
InChI Key RJPNVPITBYXBNB-UHFFFAOYSA-N
SMILES CC1=CC(F)=CC=C1Br
IUPAC Name 1-bromo-4-fluoro-2-methylbenzene
Molecular Weight (g/mol) 189.03
PubChem CID 96743
Formula Weight 189.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorotoluene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-5-fluorotoluene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-5-fluorotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.