missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzyl bromide, 97%

Product Code. 11387486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387486 1 g 1g
11397486 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387486 Supplier Thermo Scientific Alfa Aesar Supplier No. L09553.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112399-50-5 | C7H5Br2F | 267.923 g/mol

It is an important pharmaceutical intermediate. Alkylation of the β-amino ester 1a with 2-bromo-5-fluorobenzyl bromide yields Benzazepines as a white solid after silica gel chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate. Alkylation of the β-amino ester 1a with 2-bromo-5-fluorobenzyl bromide yields Benzazepines as a white solid after silica gel chromatography.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112399-50-5
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD00042187
InChI Key CZLWYKAZAVYQIK-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzyl bromide, 1-bromo-2-bromomethyl-4-fluorobenzene, 5-fluoro-2-bromobenzyl bromide, 2-bromo-5-fluorobenzylbromide, benzene, 1-bromo-2-bromomethyl-4-fluoro, alpha,2-dibromo-5-fluorotoluene, pubchem1953, acmc-1bt22, ksc493i2h, 2-bromo-5-fluoro benzyl bromide
PubChem CID 517972
IUPAC Name 1-bromo-2-(bromomethyl)-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)CBr)Br

Specifications

Melting Point 33°C to 36°C
Boiling Point 89°C to 90°C (1 mmHg)
Refractive Index 1.59
Quantity 1 g
UN Number UN3265
Beilstein 4177658
Solubility Information Difficult to mix in water.
Formula Weight 267.92
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluorobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.