missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzotrifluoride, 97%, Thermo Scientific™

Catalog Number 11904551
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
40161-55-5
C7H3BrF4
243.00
MFCD00042493
AJLIJYGWAXPEOK-UHFFFAOYSA-N
5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
2736325
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
FC1=CC=C(Br)C=C1C(F)(F)F
This item is not returnable. View return policy
40161-55-5
C7H3BrF4
243.00
MFCD00042493
AJLIJYGWAXPEOK-UHFFFAOYSA-N
5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
2736325
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
FC1=CC=C(Br)C=C1C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.7500g/mL
Boiling Point 136°C to 143°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.75
Formula Weight 243
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-fluorobenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-5-fluorobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'