missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzaldehyde, 98%

Product Code. 11466057
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466057 25 g 25g
11456057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466057 Supplier Thermo Scientific Alfa Aesar Supplier No. B25071.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94569-84-3 | C7H4BrFO | 203.01 g/mol

2-Bromo-5-fluorobenzaldehyde is used as a precursor for the synthesis of 5-fluoro-3-substituted benzoxaboroles, which is used in material science as molecular receptors, building block in crystal engineering, as steroid conjugates for molecular imprinting , dyes and biosensors of alpha hydroxyl carboxylic acids. It is also used in the synthesis of 5-arylindazolo[3,2-b]quinazolin-7(5H)-one by reacting with 2-amino-N'-arylbenzohydrazide in the presence of Copper(I) bromide by the Ullmann-type reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorobenzaldehyde is used as a precursor for the synthesis of 5-fluoro-3-substituted benzoxaboroles, which is used in material science as molecular receptors, building block in crystal engineering, as steroid conjugates for molecular imprinting , dyes and biosensors of alpha hydroxyl carboxylic acids. It is also used in the synthesis of 5-arylindazolo[3,2-b]quinazolin-7(5H)-one by reacting with 2-amino-N′-arylbenzohydrazide in the presence of Copper(I) bromide by the Ullmann-type reaction.

Solubility
Soluble in methanol.

Notes
Air sensitive. Keep container tightly closed in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94569-84-3
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00142872
InChI Key CJUCIKJLMFVWIS-UHFFFAOYSA-N
Synonym benzaldehyde, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzaldhyde, 5-fluoro-2-bromo benzaldehyde, 2-bromo-5-fluoro-benzaldehyde, pubchem1418, acmc-209rsk, ksc494g0d, 2-bromo-5-fluoro benzaldehyde, 2-bromo-5-fluorobenzaldehyde
PubChem CID 2773321
IUPAC Name 2-bromo-5-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)Br

Specifications

Melting Point 51°C to 56°C
Flash Point 87°C (190°F)
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 203.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.