missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)pyridine, 97%

Product Code. 11996471 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 11996471

Brand: Thermo Scientific Acros 445815000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 175205-81-9 | C6H3BrF3N | 226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 175205-81-9
Melting Point -21°C to 20°C
Density 1.8270g/mL
Boiling Point 161°C to 162°C
Assay Percent Range 96% min. (GC)
Molecular Formula C6H3BrF3N
Quantity 500 mg
Specific Gravity 1.827
Synonym 2-bromo-4-trifluoromethyl pyridine, pyridine, 2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl-pyridine, 2-bromo-4-trifluormethyl pyridine, pubchem8064, acmc-209e9d, abbypharma ap-11-5529
InChI Key WZVHLUMAQLUNTJ-UHFFFAOYSA-N
SMILES C1=CN=C(C=C1C(F)(F)F)Br
IUPAC Name 2-bromo-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol) 226
PubChem CID 2781543
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.