missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)pyridine, 97%

Product Code. 11319959
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11319959

Brand: Thermo Scientific Alfa Aesar H25748.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 175205-81-9 | C6H3BrF3N | 225.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175205-81-9
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD00153085
InChI Key WZVHLUMAQLUNTJ-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl pyridine, pyridine, 2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl-pyridine, 2-bromo-4-trifluormethyl pyridine, pubchem8064, acmc-209e9d, abbypharma ap-11-5529
PubChem CID 2781543
IUPAC Name 2-bromo-4-(trifluoromethyl)pyridine
SMILES C1=CN=C(C=C1C(F)(F)F)Br

Specifications

Density 1.83
Boiling Point 84°C to 85°C (14 mmHg)
Flash Point 73°C (164°F)
Refractive Index 1.478
Quantity 5 g
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.