missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-trifluoromethoxyaniline, 98%, Thermo Scientific™

Product Code. 10068851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10068851 1 g Glass bottle 1g
10500532 5 g Glass bottle 5g
10229373 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10068851 Supplier Thermo Scientific Acros Supplier No. 350170010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-17-8
Molecular Formula C7H5BrF3NO
Molecular Weight (g/mol) 256.02
MDL Number MFCD00203478
InChI Key ROSTYHNIIDIBEG-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethoxy aniline, 2-bromo-4-tirfluoromethoxyaniline, 2-bromo-4-trifluoromethoxy-phenylamine, benzenamine, 2-bromo-4-trifluoromethoxy, 2-bromo-4-trifluoromethoxy phenylamine, acmc-1c8k7, ksc495e4t, 2-bromo-4 trifluoromethoxy aniline
PubChem CID 688296
IUPAC Name 2-bromo-4-(trifluoromethoxy)aniline
SMILES NC1=CC=C(OC(F)(F)F)C=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange-Yellow to Yellow
Density 1.6930g/mL
Boiling Point 60°C to 64°C (0.3 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5030 to 1.505
Quantity 1 g
Specific Gravity 1.693
Formula Weight 256.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-trifluoromethoxyaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4-trifluoromethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.