missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-phenylacetophenone, 97%

Product Code. 11485717
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485717 100 g 100g
11465717 5 g 5g
11475717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485717 Supplier Thermo Scientific Alfa Aesar Supplier No. B24298.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135-73-9 | C14H11BrO | 275.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-73-9
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00000202
InChI Key KGHGZRVXCKCJGX-UHFFFAOYSA-N
Synonym 2-bromo-4'-phenylacetophenone, 4-phenylphenacyl bromide, p-bromoacetylbiphenyl, p-phenylphenacyl bromide, bromomethyl p-biphenylyl ketone, ethanone, 1-1,1'-biphenyl-4-yl-2-bromo, 2-bromo-1-4-phenylphenyl ethan-1-one, acetophenone, 2-bromo-4'-phenyl, alpha-bromo-p-phenylacetophenone, omega-bromo-4-phenylacetophenone
PubChem CID 67282
IUPAC Name 2-bromo-1-(4-phenylphenyl)ethanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

Melting Point 123°C to 127°C
Quantity 100 g
UN Number UN3261
Beilstein 513838
Sensitivity Light sensitive
Formula Weight 275.15
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-phenylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.