missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitrotoluene, 98%

Product Code. 11492647
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492647 25 g 25g
11402657 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492647 Supplier Thermo Scientific Alfa Aesar Supplier No. A14865.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7745-93-9 | C7H6BrNO2 | 216.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7745-93-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007195
InChI Key XFZFJQHXWJIBQV-UHFFFAOYSA-N
Synonym 2-bromo-4-nitrotoluene, benzene, 2-bromo-1-methyl-4-nitro, 3-bromo-4-methylnitrobenzene, toluene, 2-bromo-4-nitro, 3-bromo-4-methyl nitrobenzene, 1-bromo-2-methyl-5-nitrobenzene, 2-bromo-1-methyl-4-nitro-benzene, pubchem3822, 2-bromo-4nitrotoluene, 2-bromo4-nitrotoluene
PubChem CID 82189
IUPAC Name 2-bromo-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])Br

Specifications

Melting Point 74°C to 78°C
Quantity 25 g
Beilstein 1941232
Formula Weight 216.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-nitrotoluene
compliance-icons
Product Identifier
  • 2-Bromo-4-nitrotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-4-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.