missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyridine, 96%

Product Code. 11400798
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400798 1 g 1g
11410798 5 g 5g
11420798 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400798 Supplier Thermo Scientific Alfa Aesar Supplier No. B25253.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4926-28-7 | C6H6BrN | 172.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4926-28-7
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00082590
InChI Key LSZMVESSGLHDJE-UHFFFAOYSA-N
Synonym 2-bromo-4-picoline, 2-bromo-4-methyl-pyridine, 4-methyl-2-bromopyridine, pyridine, 2-bromo-4-methyl, 2 bromo-4-methyl pyridine, 2-bromo-4-methyl pyridine, pubchem1184, acmc-209kfi, 2-bromo4-methylpyridine
PubChem CID 2734087
IUPAC Name 2-bromo-4-methylpyridine
SMILES CC1=CC=NC(Br)=C1

Specifications

Density 1.545
Boiling Point 87°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 1 g
Beilstein 107331
Formula Weight 172.03
Percent Purity 96%
Chemical Name or Material 2-Bromo-4-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.