missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methylpropiophenone, 98%

Product Code. 15462279
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462279 25 g 25g
15442279 1 g 1g
15452279 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462279 Supplier Thermo Scientific Alfa Aesar Supplier No. H64881.14
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1451-82-7 | C10H11BrO | 227.10 g/mol

It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

Solubility
Soluble in chloroform and methanol.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1451-82-7
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.10
MDL Number MFCD11131402
InChI Key OZLUPIIIHOOPNQ-UHFFFAOYNA-N
Synonym 2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone
PubChem CID 2734063
IUPAC Name 2-bromo-1-(4-methylphenyl)propan-1-one
SMILES CC(Br)C(=O)C1=CC=C(C)C=C1

Specifications

Quantity 25 g
Beilstein 2207628
Solubility Information Soluble in chloroform and methanol.
Formula Weight 227.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4'-methylpropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.