missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylphenol, 97%

Product Code. 11410297
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410297 5 g 5g
11420297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410297 Supplier Thermo Scientific Alfa Aesar Supplier No. B21701.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6627-55-0 | C7H7BrO | 187.036 g/mol

It is applied as a compound responsible for iodine off-flavor in wines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a compound responsible for iodine off-flavor in wines.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6627-55-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00002151
InChI Key MTIDYGLTAOZOGU-UHFFFAOYSA-N
Synonym 2-bromo-p-cresol, phenol, 2-bromo-4-methyl, 3-bromo-4-hydroxytoluene, 2-bromo-4-methyl-phenol, p-cresol, 2-bromo, pubchem4096, acmc-209nul, 4-06-00-02143 beilstein handbook reference, ksc494c6f
PubChem CID 23109
IUPAC Name 2-bromo-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)Br

Specifications

Melting Point 16°C
Density 1.547
Boiling Point 213°C to 214°C
Flash Point >110°C (230°F)
Odor Phenol-like
Refractive Index 1.578
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4-methylpheno
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-p-cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.