missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylbenzoic acid, 97%

Product Code. 15466878
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15466878

Brand: Thermo Scientific Alfa Aesar H60413.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7697-27-0 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-27-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00045797
InChI Key ZZYYOHPHSYCHQG-UHFFFAOYSA-N
Synonym 4-methyl-2-bromobenzoic acid, 2-bromo-p-toluic acid, 2-bromo-4-methyl-benzoic acid, benzoic acid, 2-bromo-4-methyl, 2-bromo-4-methyl benzoic acid, zlchem 427, pubchem4719, 3-bromo-4-carboxytoluene, acmc-1bbb4, 2-bromo-4-methylbenzoicacid
PubChem CID 226292
IUPAC Name 2-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(Br)=C1

Specifications

Melting Point 143°C to 147°C
Quantity 1 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.