missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylaniline, 99%

Product Code. 11414904
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414904 50 g 50g
11424904 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414904 Supplier Thermo Scientific Alfa Aesar Supplier No. A13818.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 583-68-6 | C7H8BrN | 186.052 g/mol

The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-68-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00007635
InChI Key UVRRJILIXQAAFK-UHFFFAOYSA-N
Synonym 2-bromo-p-toluidine, benzenamine, 2-bromo-4-methyl, 4-amino-3-bromotoluene, 2-bromo-4-methylbenzenamine, p-toluidine, 2-bromo, 2-bromo-4-methyl-phenylamine, 3-bromo-4-aminotoluene, 4-methyl-2-bromoaniline, 2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline
PubChem CID 11422
IUPAC Name 2-bromo-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Br

Specifications

Melting Point 14°C to 16°C
Density 1.494
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.603
Quantity 50 g
UN Number UN2810
Beilstein 1931711
Solubility Information Insoluble in water.
Formula Weight 186.06
Percent Purity 99%
Chemical Name or Material 2-Bromo-4-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.