missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methylacetophenone, 97%

Product Code. 10736622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10736622 25 g Plastic bottle 25g
10193571 5 g Glass bottle 5g
10464735 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10736622 Supplier Thermo Scientific Acros Supplier No. 170390250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-41-0 | C9H9BrO | 213.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-41-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00000203
InChI Key KRVGXFREOJHJAX-UHFFFAOYSA-N
Synonym 2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
PubChem CID 69272
IUPAC Name 2-bromo-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 52°C to 53°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C6H4COCH2Br
Quantity 25 g
Beilstein 07, 309
Formula Weight 213.07
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2-Bromo-4'-methylacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4'-methylacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.