missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methoxyacetophenone, TRC

Product Code. 30342133
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342133 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342133 Supplier Toronto Research Chemicals Supplier No. TRCB68472025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-4'-methoxyacetophenone
CAS 2632-13-5
Molecular Formula C9H9BrO2
Quantity 25 g
Synonym 2-Bromo-1-(4-methoxyphenyl)ethanone, 2-(4-Methoxyphenyl)-2-oxoethyl bromide, 2-Bromo-1-(4-methoxyphenyl)-1-ethanone, 2-Bromo-1-[4-(methyloxy)phenyl]ethanone, 2-Bromo-p-methoxyacetophenone, 4-Methoxy-2'-bromoacetophenone, 4-Methoxyphenacyl Bromide, 4'-Methoxyphenacyl Bromide, Bromomethyl 4-Methoxyphenyl Ketone, Bromomethyl p-Anisyl Ketone, Bromomethyl p-Methoxyphenyl Ketone, NSC 129010, p-Methoxyphenacyl Bromide, alpha-Bromo-4'-methoxyacetophenone, alpha-Bromo-p-methoxyacetophenone, ω-Bromo-4'-methoxyacetophenone, ω-Bromo-p-methoxyacetophenone
InChI Formula InChI=1S/C9H9BrO2/c1-12-8-4-2-7(3-5-8)9(11)6-10/h2-5H,6H2,1H3
SMILES COc1ccc(cc1)C(=O)CBr
IUPAC Name 2-bromo-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol) 229.07
Formula Weight 227.9786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.