missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methoxyacetophenone, 98%

Product Code. 11425443
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425443 5 g 5g
11435443 25 g 25g
11445443 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425443 Supplier Thermo Scientific Alfa Aesar Supplier No. A13415.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2632-13-5 | C9H9BrO2 | 229.07 g/mol

2-Bromo-4'-methoxyacetophenone, is used as a cell-permeable, covalent and potent protein tyrosine phosphatase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-methoxyacetophenone, is used as a cell-permeable, covalent and potent protein tyrosine phosphatase inhibitor.

Solubility
It is soluble in DMSO, water (partly miscible), most organic solvents, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2632-13-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000201
InChI Key XQJAHBHCLXUGEP-UHFFFAOYSA-N
Synonym 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
PubChem CID 4965
IUPAC Name 2-bromo-1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 68°C to 72°C
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 743112
Solubility Information It is soluble in DMSO,water (partly miscible),most organic solvents,and methanol.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-methoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.