missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methoxyacetophenone, 98%

Product Code. 10143200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143200 5 g Glass bottle 5g
10531401 25 g Glass bottle 25g
10300404 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143200 Supplier Thermo Scientific Acros Supplier No. 151900050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2632-13-5 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2632-13-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000201
InChI Key XQJAHBHCLXUGEP-UHFFFAOYSA-N
Synonym 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
PubChem CID 4965
IUPAC Name 2-bromo-1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 72°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCH2Br
Quantity 5 g
Beilstein 08, 89
Fieser 05,439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 229.07
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromo-4'-methoxyacetophenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-4'-methoxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.